Assay Details
| Gene Symbol: E430025E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153548.2 | NP_705776.2 | 17 - 18 | 2334 | 85 | |
| GenBank mRNA | AK128970.1 | - | 17 - 18 | 2349 | 85 | |
| AK129086.1 | - | 63 | 85 | |||
| BC031364.1 | - | 3 - 4 | 348 | 85 | ||
| BC034070.1 | - | 7 - 8 | 934 | 4217923 | 85 | |
| BC040815.1 | - | 7 - 8 | 934 | 4217923 | 85 | |
| BC067035.1 | - | 17 - 18 | 2334 | 30532658 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

E430025E21Rik
Genomic Map