Assay Details
| Gene Symbol: E430025E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153548.2 | NP_705776.2 | 21 - 22 | 2899 | 118 | |
| GenBank mRNA | AK088754.1 | - | 6 - 7 | 772 | 118 | |
| AK128970.1 | - | 21 - 22 | 2914 | 118 | ||
| AK129086.1 | - | 2551 | 118 | |||
| AK163968.1 | - | 4 - 5 | 523 | 118 | ||
| BC031364.1 | - | 7 - 8 | 913 | 118 | ||
| BC034070.1 | - | 11 - 12 | 1499 | 4217923 | 118 | |
| BC040815.1 | - | 11 - 12 | 1499 | 4217923 | 118 | |
| BC067035.1 | - | 21 - 22 | 2899 | 30532658 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

E430025E21Rik
Genomic Map