Assay Details
| Gene Symbol: E430025E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153548.2 | NP_705776.2 | 23 - 24 | 3083 | 90 | |
| GenBank mRNA | AK088754.1 | - | 8 - 9 | 956 | 90 | |
| AK129086.1 | - | 2735 | 90 | |||
| AK163968.1 | - | 6 - 7 | 707 | 90 | ||
| BC031364.1 | - | 9 - 10 | 1097 | 90 | ||
| BC034070.1 | - | 13 - 14 | 1683 | 4217923 | 90 | |
| BC040815.1 | - | 13 - 14 | 1683 | 4217923 | 90 | |
| BC067035.1 | - | 23 - 24 | 3083 | 30532658 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

E430025E21Rik
Genomic Map