Assay Details
| Gene Symbol: E430025E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153548.2 | NP_705776.2 | 7 - 8 | 1093 | 108 | |
| GenBank mRNA | AK128970.1 | - | 7 - 8 | 1108 | 108 | |
| BC067035.1 | - | 7 - 8 | 1093 | 30532658 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

E430025E21Rik
Genomic Map