Assay Details
| Gene Symbol: Myrf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033481.1 | NP_001028653.1 | 7 - 8 | 1257 | 60 | |
| GenBank mRNA | AK141695.1 | - | 7 - 8 | 1258 | 60 | |
| BC157942.1 | - | 4 - 5 | 648 | 9007277 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
proteolysis
oligodendrocyte development
central nervous system myelination
cell differentiation
positive regulation of myelination
central nervous system myelin maintenance
positive regulation of transcription, DNA-templated
oligodendrocyte differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
peptidase activity
cysteine-type peptidase activity
hydrolase activity
sequence-specific DNA binding
transcription, DNA-templated
regulation of transcription, DNA-templated
proteolysis
oligodendrocyte development
central nervous system myelination
cell differentiation
positive regulation of myelination
central nervous system myelin maintenance
positive regulation of transcription, DNA-templated
oligodendrocyte differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
peptidase activity
cysteine-type peptidase activity
hydrolase activity
sequence-specific DNA binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P104446 | BC157942.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P104446 | AK039649.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P104446 | NM_001033481.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206154 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 104623 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 657005 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149273 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568404 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 132525 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 111823 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 863918 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955552 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 664167 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 184749 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 72289 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 27709 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 57577 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 11771 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342951 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 160354 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 463204 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494989 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877606 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 66186 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 99792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 133873 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P104446 | BC157942.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P104446 | AK039649.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P104446 | NM_001033481.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P457975 | AK031485.1 | mouse_transcriptome |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10465939 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465940 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465923 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465925 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465928 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465933 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465941 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465921 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465931 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465929 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465920 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465937 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465926 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465938 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465934 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465927 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465918 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465932 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465930 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465917 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465935 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465942 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465924 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465936 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465922 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465919 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439506_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10465916 | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 225908_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 225908_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900010258.mm.1 | NM_001033481.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010258.mm.1 | NM_001033481.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010258.mm.1 | TC1900001097.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 104266_PM_at | - | - |
| Murine Genome U74v2 Set | 104266_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439506_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439506_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2426233 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1248687 | - | - |

Myrf
Genomic Map