Assay Details
| Gene Symbol: Irf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008391.4 | NP_032417.3 | 1 - 2 | 192 | 82 | |
| GenBank mRNA | AK036652.1 | - | 1 - 2 | 160 | 82 | |
| AK087926.1 | - | 1 - 2 | 178 | 82 | ||
| AK132099.1 | - | 1 - 2 | 205 | 82 | ||
| AK155025.1 | - | 1 - 2 | 177 | 82 | ||
| BC006577.1 | - | 1 - 2 | 185 | 3586586 | 81 | |
| BC110666.1 | - | 1 - 2 | 164 | 4918447 | 82 | |
| J03168.1 | - | 1 - 2 | 147 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK155025.1, BC006577.1, BC110666.1, J03168.1, NM_008391.4 transcripts.
Set Membership: |
|

Irf2
Genomic Map