Assay Details
| Gene Symbol: Adrbk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285806.1 | NP_001272735.1 | 14 - 15 | 1320 | 63 | |
| NM_177078.4 | NP_796052.2 | 14 - 15 | 1281 | 63 | ||
| GenBank mRNA | AK076533.1 | - | 14 - 15 | 1179 | 63 | |
| AK134672.1 | - | 14 - 15 | 1258 | 63 | ||
| AK141186.1 | - | 14 - 15 | 1300 | 63 | ||
More Information
Set Membership: |
|

Adrbk2
Genomic Map