Assay Details
| Gene Symbol: Pcgf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027654.2 | NP_081930.1 | 5 - 6 | 711 | 157 | |
| GenBank mRNA | AB047007.1 | - | 5 - 6 | 761 | 157 | |
| AK016709.1 | - | 5 - 6 | 158 | |||
| AK132448.1 | - | 5 - 6 | 757 | 157 | ||
| BC016195.1 | - | 5 - 6 | 711 | 4488614 | 157 | |
| BC089460.1 | - | 5 - 6 | 750 | 30632537 | 157 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcgf6
Genomic Map