Assay Details
| Gene Symbol: Abhd17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146096.3 | NP_666208.2 | 1 - 2 | 285 | 148 | |
| GenBank mRNA | AK050879.1 | - | 1 - 2 | 410 | 148 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_146096.3 transcripts.
Set Membership: |
|

Abhd17b
Genomic Map