Assay Details
| Gene Symbol: Cyb561d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081320.2 | NP_001074789.2 | 2 - 3 | 247 | 57 | |
| NM_028061.1 | NP_082337.1 | 2 - 3 | 114 | 57 | ||
| GenBank mRNA | AK005419.1 | - | 1 - 2 | 314 | 57 | |
| AK087683.1 | - | 2 - 3 | 194 | 57 | ||
| AK164441.1 | - | 2 - 3 | 121 | 57 | ||
| BB853561.1 | - | 2 - 3 | 247 | 57 | ||
| BC043107.1 | - | 2 - 3 | 733 | 57 | ||
| BC060107.1 | - | 2 - 3 | 89 | 57 | ||
| BC137590.1 | - | 2 - 3 | 224 | 8860609 | 57 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK164441.1, NM_028061.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P352283 | AK087683.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P352283 | AK087683.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071087 | BC137590.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071087 | NM_001081320.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071087 | NM_028061.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071087 | AK005419.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071087 | AK087683.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 860333 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 685174 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316445 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105585 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P352283 | AK087683.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P352283 | AK087683.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071087 | BC137590.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071087 | NM_001081320.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071087 | NM_028061.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071087 | AK005419.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2071087 | AK087683.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105585 | - | - |
| Whole Mouse Genome G4122A | A_51_P352283 | AK087683.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P352283 | AK087683.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125834 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22411269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02757395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02621631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02621616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02879891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02621622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411204 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03063343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02511995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02621619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411148 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02621609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411199 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411193 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22411175 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10501284 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501285 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501283 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434226_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431021_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10501282 | - | - |
| Mouse Genome 430 2.0 Array | 1459837_at | - | - |
| Mouse Genome 430 2.0 Array | 1459837_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431021_at | - | - |
| Mouse Genome 430 2.0 Array | 1434226_at | - | - |
| Mouse Genome 430 2.0 Array | 1431021_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434226_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72023_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72023_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300021432.mm.1 | NM_001081320.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021432.mm.1 | NM_001081320.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021432.mm.1 | NM_028061.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021432.mm.1 | NM_028061.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021432.mm.1 | TC0300002734.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021434.mm.1 | NM_001081320.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021434.mm.1 | NM_001081320.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021434.mm.1 | NM_028061.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021434.mm.1 | NM_028061.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021434.mm.1 | TC0300002734.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021440.mm.1 | NM_001081320.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021440.mm.1 | NM_001081320.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021440.mm.1 | NM_028061.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021440.mm.1 | NM_028061.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021440.mm.1 | TC0300002734.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021433.mm.1 | NM_001081320.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021433.mm.1 | NM_001081320.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021433.mm.1 | NM_028061.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021433.mm.1 | NM_028061.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021433.mm.1 | TC0300002734.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434226_PM_at | - | - |
| Mouse Expression Set 430 | 1434226_at | - | - |
| Mouse Expression Set 430 | 1431021_PM_at | - | - |
| Mouse Expression Set 430 | 1431021_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434226_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431021_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431021_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434226_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 768008 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103060053 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103060053 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225616 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2624360 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103060053 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103060053 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3078306 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2624358 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3078306 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225616 | - | - |

Cyb561d1
Genomic Map