Assay Details
| Gene Symbol: Pkd2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181422.3 | NP_852087.2 | 9 - 10 | 1825 | 75 | |
| GenBank mRNA | AB290927.1 | - | 9 - 10 | 1657 | 75 | |
| AF271381.1 | - | 9 - 10 | 1791 | 75 | ||
| AK046772.1 | - | 9 - 10 | 1826 | 75 | ||
| BC046386.1 | - | 4 - 5 | 518 | 75 | ||
| BC116297.2 | - | 9 - 10 | 1756 | 40094997 | 75 | |
| BC116323.2 | - | 9 - 10 | 1756 | 40096335 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pkd2l1
Genomic Map