Assay Details
| Gene Symbol: Klk13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039042.2 | NP_001034131.2 | 5 - 6 | 827 | 89 | |
| GenBank mRNA | BC063763.1 | - | 5 - 6 | 828 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084965 | BC063763.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084965 | NM_001039042.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 667823 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 272663 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 447619 | AK006852.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552891 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 583765 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 957588 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 261726 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604364 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 38413 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 8040 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159406 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147474 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084965 | BC063763.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084965 | NM_001039042.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083748 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083725 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083734 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25083758 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01505146 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06178184 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147474 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159406 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147474 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25083728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06328805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06016716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06167441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06103876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06421638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06234699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06384059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06450289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06234700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06286419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06123950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06328809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06167438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25083732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06488423 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10552474 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552473 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552472 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552470 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552471 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10552469 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700006254.mm.1 | NM_001039042.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006254.mm.1 | NM_001039042.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006254.mm.1 | TC0700000770.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700006255.mm.1 | NM_001039042.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006255.mm.1 | NM_001039042.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006255.mm.1 | TC0700000770.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237292 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3115315 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3040588 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3040588 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3115315 | - | - |

Klk13
Genomic Map