Assay Details
| Gene Symbol: Lrrc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198119.2 | NP_932787.1 | 3 - 4 | 430 | 88 | |
| GenBank mRNA | AK048678.1 | - | 3 - 4 | 423 | 88 | |
| AK078176.1 | - | 3 - 4 | 431 | 88 | ||
| BC116884.1 | - | 1 - 2 | 222 | 40126203 | 88 | |
| BC116886.1 | - | 1 - 2 | 222 | 40126205 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc24
Genomic Map