Assay Details
| Gene Symbol: Lrrc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198119.2 | NP_932787.1 | 4 - 5 | 707 | 81 | |
| GenBank mRNA | AK048678.1 | - | 4 - 5 | 700 | 81 | |
| AK078176.1 | - | 4 - 5 | 708 | 81 | ||
| BC116884.1 | - | 2 - 3 | 499 | 40126203 | 81 | |
| BC116886.1 | - | 2 - 3 | 499 | 40126205 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc24
Genomic Map