Assay Details
| Gene Symbol: Lrrc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198119.2 | NP_932787.1 | 5 - 6 | 871 | 79 | |
| GenBank mRNA | AK048678.1 | - | 5 - 6 | 864 | 79 | |
| AK078176.1 | - | 5 - 6 | 872 | 79 | ||
| BC116884.1 | - | 3 - 4 | 663 | 40126203 | 79 | |
| BC116886.1 | - | 3 - 4 | 663 | 40126205 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350165 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350214 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350215 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350212 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350168 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350213 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350220 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350222 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350167 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350211 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P11402 | BC116884.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P11402 | BC116886.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P11402 | NM_198119.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P11402 | AK048678.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P11402 | AK078176.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P11402 | BC116884.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P11402 | BC116886.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P11402 | NM_198119.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P11402 | AK048678.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P11402 | AK078176.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 202502 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 864815 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868056 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 460962 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127292 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P11402 | BC116884.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P11402 | BC116886.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P11402 | NM_198119.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P11402 | AK048678.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P11402 | AK078176.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P11402 | BC116884.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P11402 | BC116886.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P11402 | NM_198119.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P11402 | AK048678.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P11402 | AK078176.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30350220 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127292 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08423229 | - | - |
| Whole Mouse Genome G4122A | A_52_P11402 | BC116884.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P11402 | BC116886.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P11402 | NM_198119.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P11402 | AK048678.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P11402 | AK078176.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P11402 | BC116884.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P11402 | BC116886.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P11402 | NM_198119.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P11402 | AK048678.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P11402 | AK078176.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127292 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12180191 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12180213 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12180199 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12180200 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350215 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350211 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350168 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350165 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350220 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350213 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350214 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350222 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350167 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30350212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30350126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12180200 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350200 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12319841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350202 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12180212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350176 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12319809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12333820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12180210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350174 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12028701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12180198 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350208 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30350212 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10430103 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430102 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430101 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430098 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430107 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430097 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430099 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430104 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10430100 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433876_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10430096 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10430105 | - | - |
| Mouse Genome 430 2.0 Array | 1433876_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1433876_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 378937_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 378937_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500013888.mm.1 | NM_198119.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500013888.mm.1 | NM_198119.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500013888.mm.1 | TC1500001790.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500013890.mm.1 | NM_198119.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500013890.mm.1 | NM_198119.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500013890.mm.1 | TC1500001790.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500013889.mm.1 | NM_198119.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500013889.mm.1 | NM_198119.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500013889.mm.1 | TC1500001790.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1433876_PM_at | - | - |
| Mouse Expression Set 430 | 1433876_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433876_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433876_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6650193 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006650193 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2684397 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2684397 | - | - |
| mouse-6 v1.1 (Illumina) | scl0378937.1_229-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6650193 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0378937.1_229-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6650193 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2684397 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2684397 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0378937.1_229-S | - | - |

Lrrc24
Genomic Map