Assay Details
| Gene Symbol: Sumf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026445.2 | NP_080721.1 | 7 - 8 | 703 | 69 | |
| GenBank mRNA | AK013017.1 | - | 7 - 8 | 683 | 69 | |
| AK076022.1 | - | 7 - 8 | 709 | 69 | ||
| AK138300.1 | - | 7 - 8 | 694 | 69 | ||
| AK140169.1 | - | 8 - 9 | 718 | 69 | ||
| AK157784.1 | - | 7 - 8 | 704 | 69 | ||
| AK166216.1 | - | 7 - 8 | 682 | 69 | ||
| BC119480.2 | - | 5 - 6 | 812 | 40050522 | 69 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974442 | AK157784.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974442 | AK138300.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974442 | AK140169.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974442 | NM_026445.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162835 | AK157784.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162835 | AK138300.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162835 | NM_026445.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162835 | BC055102.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162835 | J03293.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 951238 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923634 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715144 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75220 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 37776 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 245058 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 811426 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 505387 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815149 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340740 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 709521 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 483850 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 194047 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924137 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 871253 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 125198 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 80355 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 151357 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 69904 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 77738 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 38133 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126671 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158414 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974442 | AK157784.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974442 | AK138300.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974442 | AK140169.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974442 | NM_026445.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162835 | AK157784.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162835 | AK138300.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162835 | NM_026445.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162835 | BC055102.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162835 | J03293.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090894 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126671 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01220262 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158414 | - | - |
| Whole Mouse Genome G4122A | A_51_P321825 | AK157784.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321825 | AK215029.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321825 | AK138300.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321825 | AK140169.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321825 | AK076022.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321820 | AK157784.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321820 | AK138300.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321820 | AK140169.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321820 | AK076022.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321820 | NM_026445.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126671 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24091010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24091012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24091001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24091003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24091007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090901 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090944 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04680197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04646703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090931 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04377460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04878348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090910 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10526093 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526091 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526088 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526089 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526090 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526094 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526096 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526095 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526092 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526097 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429713_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10526087 | - | - |
| Mouse Genome 430 2.0 Array | 1429713_at | - | - |
| Mouse Genome 430 2.0 Array | 1429713_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67902_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67902_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500013699.mm.1 | NM_026445.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013699.mm.1 | NM_026445.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013699.mm.1 | TC0500001514.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500013698.mm.1 | NM_026445.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013698.mm.1 | NM_026445.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013698.mm.1 | TC0500001514.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500013700.mm.1 | NM_026445.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013700.mm.1 | NM_026445.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013700.mm.1 | TC0500001514.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500013701.mm.1 | NM_026445.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013701.mm.1 | NM_026445.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013701.mm.1 | TC0500001514.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429713_PM_at | - | - |
| Mouse Expression Set 430 | 1429714_PM_at | - | - |
| Mouse Expression Set 430 | 1429714_at | - | - |
| Mouse Expression Set 430 | 1429713_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429713_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429713_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 786790 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104610102 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1239103 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2764127 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2534816 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3040485 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3115185 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3040485 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3115185 | - | - |

Sumf2
Genomic Map