Assay Details
| Gene Symbol: Asl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133768.4 | NP_598529.1 | 14 - 15 | 1140 | 60 | |
| GenBank mRNA | AK010922.1 | - | 14 - 15 | 1152 | 60 | |
| AK076453.1 | - | 14 - 15 | 1155 | 60 | ||
| AK138423.1 | - | 14 - 15 | 1137 | 60 | ||
| AK140160.1 | - | 14 - 15 | 1155 | 60 | ||
| AK148764.1 | - | 14 - 15 | 1124 | 60 | ||
| AK168739.1 | - | 14 - 15 | 1135 | 60 | ||
| AK186697.1 | - | 2 - 3 | 136 | 60 | ||
| AK215777.1 | - | 2 - 3 | 136 | 60 | ||
| BC016670.1 | - | 14 - 15 | 1140 | 2648753 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asl
Genomic Map