| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800013365.mm.1 |
NM_001037800.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013365.mm.1 |
NM_001287211.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013365.mm.1 |
NM_026972.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013365.mm.1 |
NR_104430.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013365.mm.1 |
TC0800001624.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800013366.mm.1 |
NM_001037800.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013366.mm.1 |
NM_001287211.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013366.mm.1 |
NM_026972.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013366.mm.1 |
NR_104430.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013366.mm.1 |
TC0800001624.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800013369.mm.1 |
NM_001037800.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013369.mm.1 |
NM_001287211.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013369.mm.1 |
NM_026972.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013369.mm.1 |
NR_104430.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800013369.mm.1 |
TC0800001624.mm.1 |
MTA TC |