Assay Details
| Gene Symbol: Cd209b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037800.3 | NP_001032889.3 | 7 - 8 | 673 | 88 | |
| NM_001287211.1 | NP_001274140.1 | 7 - 8 | 673 | 88 | ||
| NM_026972.5 | NP_081248.4 | 8 - 9 | 763 | 88 | ||
| GenBank mRNA | AF373409.1 | - | 8 - 9 | 741 | 88 | |
| AF374471.1 | - | 7 - 8 | 630 | 88 | ||
| AF422108.1 | - | 8 - 9 | 736 | 88 | ||
| AF424790.1 | - | 8 - 9 | 735 | 88 | ||
| AF424791.1 | - | 8 - 9 | 735 | 88 | ||
| AF424792.1 | - | 8 - 9 | 88 | |||
| AF424795.1 | - | 8 - 9 | 752 | 88 | ||
| AF424796.1 | - | 8 - 9 | 746 | 88 | ||
| AF424797.1 | - | 7 - 8 | 645 | 88 | ||
| AF424798.1 | - | 7 - 8 | 665 | 88 | ||
| AF424799.1 | - | 7 - 8 | 645 | 88 | ||
| AF424800.1 | - | 2 - 3 | 166 | 88 | ||
| AF424801.1 | - | 2 - 3 | 166 | 88 | ||
| AF424802.1 | - | 2 - 3 | 166 | 88 | ||
| AK007656.1 | - | 7 - 8 | 679 | 88 | ||
| AK138999.1 | - | 8 - 9 | 737 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd209b
Genomic Map