Assay Details
| Gene Symbol: Ubc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019639.4 | NP_062613.3 | 1 - 2 | 104 | 80 | |
| GenBank mRNA | CF172065.1 | - | 1 - 2 | 104 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_019639.4 transcripts.
Set Membership: |
|

Ubc
Genomic Map