Assay Details
| Gene Symbol: Hif1an | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176958.3 | NP_795932.2 | 1 - 2 | 201 | 72 | |
| GenBank mRNA | AK043636.1 | - | 1 - 2 | 187 | 72 | |
| AK084960.1 | - | 1 - 2 | 196 | 72 | ||
| AK141867.1 | - | 1 - 2 | 187 | 72 | ||
| AK154774.1 | - | 1 - 2 | 201 | 72 | ||
| BC130013.1 | - | 1 - 2 | 205 | 40140349 | 72 | |
| BY122345.1 | - | 1 - 2 | 205 | 72 | ||
| BY713373.1 | - | 1 - 2 | 187 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hif1an
Genomic Map