Assay Details
| Gene Symbol: Hif1an | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176958.3 | NP_795932.2 | 6 - 7 | 918 | 124 | |
| GenBank mRNA | AK154774.1 | - | 6 - 7 | 918 | 124 | |
| BC130013.1 | - | 6 - 7 | 922 | 40140349 | 124 | |
| CN535711.1 | - | 5 - 6 | 502 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hif1an
Genomic Map