Assay Details
| Gene Symbol: Btrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037758.2 | NP_001032847.2 | 6 - 7 | 849 | 90 | |
| NM_001286465.1 | NP_001273394.1 | 6 - 7 | 849 | 90 | ||
| NM_001286466.1 | NP_001273395.1 | 5 - 6 | 617 | 90 | ||
| NM_009771.3 | NP_033901.1 | 5 - 6 | 741 | 90 | ||
| GenBank mRNA | AF081887.1 | - | 5 - 6 | 726 | 90 | |
| AF099932.1 | - | 5 - 6 | 636 | 90 | ||
| AF110396.1 | - | 5 - 6 | 636 | 90 | ||
| AF112979.1 | - | 5 - 6 | 90 | |||
| AK032221.1 | - | 6 - 7 | 812 | 90 | ||
| AK052317.1 | - | 7 - 8 | 873 | 90 | ||
| AK083257.1 | - | 6 - 7 | 825 | 90 | ||
| AK145091.1 | - | 5 - 6 | 553 | 90 | ||
| AK145624.1 | - | 6 - 7 | 832 | 90 | ||
| AK156660.1 | - | 5 - 6 | 90 | |||
| AK220183.1 | - | 6 - 7 | 90 | |||
| BC003989.1 | - | 5 - 6 | 642 | 3491843 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Btrc
Genomic Map