Assay Details
| Gene Symbol: Slc7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044740.2 | NP_001038205.1 | 2 - 3 | 583 | 70 | |
| NM_007514.3 | NP_031540.2 | 2 - 3 | 583 | 70 | ||
| GenBank mRNA | AK154621.1 | - | 2 - 3 | 557 | 70 | |
| AK155558.1 | - | 2 - 3 | 558 | 70 | ||
| AK171369.1 | - | 2 - 3 | 555 | 70 | ||
| BC021653.1 | - | 2 - 3 | 577 | 70 | ||
| BC127082.1 | - | 2 - 3 | 499 | 40111372 | 70 | |
| DQ086834.1 | - | 2 - 3 | 538 | 70 | ||
| L03290.1 | - | 1 - 2 | 385 | 70 | ||
| L11600.1 | - | 1 - 2 | 385 | 70 | ||
| L29006.1 | - | 1 - 2 | 908 | 70 | ||
| M62838.1 | - | 2 - 3 | 496 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a2
Genomic Map