Assay Details
| Gene Symbol: Slc7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007514.3 | NP_031540.2 | 6 - 7 | 1266 | 74 | |
| GenBank mRNA | BC021653.1 | - | 6 - 7 | 1261 | 74 | |
| BC127082.1 | - | 6 - 7 | 1182 | 40111372 | 74 | |
| DQ086834.1 | - | 6 - 7 | 1221 | 74 | ||
| L03290.1 | - | 5 - 6 | 1068 | 74 | ||
| L11600.1 | - | 5 - 6 | 1068 | 74 | ||
| L29006.1 | - | 5 - 6 | 1591 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a2
Genomic Map