Assay Details
| Gene Symbol: Rilpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030259.1 | NP_084535.1 | 2 - 3 | 608 | 66 | |
| GenBank mRNA | AK133760.1 | - | 2 - 3 | 740 | 66 | |
| AK150123.1 | - | 2 - 3 | 712 | 66 | ||
| AK150252.1 | - | 2 - 3 | 711 | 66 | ||
| BC003324.1 | - | 2 - 3 | 608 | 3156139 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rilpl2
Genomic Map