Assay Details
| Gene Symbol: Rilpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030259.1 | NP_084535.1 | 3 - 4 | 715 | 81 | |
| GenBank mRNA | AK133760.1 | - | 3 - 4 | 847 | 81 | |
| AK150123.1 | - | 3 - 4 | 819 | 81 | ||
| AK150252.1 | - | 3 - 4 | 818 | 81 | ||
| BC003324.1 | - | 3 - 4 | 715 | 3156139 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24061485 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24061481 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24061483 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24061484 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24061482 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344399 | BC003324.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344399 | NM_030259.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344399 | AK133760.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344399 | AK150123.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344399 | AK150252.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344399 | BC003324.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344399 | NM_030259.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344399 | AK133760.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344399 | AK150123.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344399 | AK150252.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313803 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 87342 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 74521 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655366 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703278 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 187009 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 528608 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 279408 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 68991 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 298674 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 106585 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114671 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164199 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344399 | BC003324.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344399 | NM_030259.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344399 | AK133760.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344399 | AK150123.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344399 | AK150252.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344399 | BC003324.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344399 | NM_030259.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344399 | AK133760.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344399 | AK150123.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344399 | AK150252.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24061501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24061506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24061496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24061485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24061486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24061493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24061480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24061478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24061491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24061498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24061503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24061482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24061481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24061476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24061477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24061489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24061474 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24061505 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05730033 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05730021 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114671 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164199 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05730014 | - | - |
| Whole Mouse Genome G4122A | A_51_P344399 | BC003324.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344399 | NM_030259.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344399 | AK133760.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344399 | AK150123.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344399 | AK150252.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344399 | BC003324.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344399 | NM_030259.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344399 | AK133760.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344399 | AK150123.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P344399 | AK150252.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107490 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111275 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114671 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164199 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05730014 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04544870 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04116803 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04544863 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04116808 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04544869 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04621660 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24061484 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24061481 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24061485 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24061482 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24061483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24061464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061496 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04143176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04832754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04143181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04800635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04114872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061467 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061454 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04544864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24061457 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10533847 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533848 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533845 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533846 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423134_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10533844 | - | - |
| Mouse Genome 430 2.0 Array | 1438403_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1446055_at | - | - |
| Mouse Genome 430 2.0 Array | 1446055_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423134_at | - | - |
| Mouse Genome 430 2.0 Array | 1423134_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438403_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 80291_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 80291_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500029441.mm.1 | NM_030259.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500029441.mm.1 | NM_030259.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500029441.mm.1 | TC0500003256.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423134_PM_at | - | - |
| Mouse Expression Set 430 | 1423134_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423134_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423134_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423134_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423134_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 558198 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1050435 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1050435 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001050435 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2777972 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2777972 | - | - |
| mouse-6 v1.1 (Illumina) | scl26031.4.1_17-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1050435 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl26031.4.1_17-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1050435 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2833985 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2833985 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2763710 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl26031.4.1_17-S | - | - |

Rilpl2
Genomic Map