Assay Details
| Gene Symbol: Cept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293694.1 | NP_001280623.1 | 1 - 2 | 126 | 108 | |
| NM_133869.4 | NP_598630.2 | 1 - 2 | 126 | 108 | ||
| GenBank mRNA | AK037116.1 | - | 1 - 2 | 86 | 108 | |
| BC021753.1 | - | 1 - 2 | 55 | 4167222 | 108 | |
| BC023783.1 | - | 1 - 2 | 66 | 5346896 | 108 | |
| BY226430.1 | - | 1 - 2 | 128 | 108 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK037116.1, BC021753.1, BC023783.1, NM_001293694.1, NM_133869.4 transcripts.
Set Membership: |
|

Cept1
Genomic Map