Assay Details
| Gene Symbol: Cept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293694.1 | NP_001280623.1 | 6 - 7 | 1048 | 124 | |
| NM_133869.4 | NP_598630.2 | 6 - 7 | 1048 | 124 | ||
| GenBank mRNA | AK037116.1 | - | 6 - 7 | 1008 | 124 | |
| BC021753.1 | - | 6 - 7 | 977 | 4167222 | 124 | |
| BC023783.1 | - | 6 - 7 | 988 | 5346896 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cept1
Genomic Map