Assay Details
| Gene Symbol: Ntng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163350.1 | NP_001156822.1 | 5 - 6 | 2188 | 102 | |
| GenBank mRNA | AB038664.1 | - | 4 - 5 | 1093 | 102 | |
| AF475072.1 | - | 4 - 5 | 1093 | 102 | ||
More Information
Set Membership: |
|

Ntng1
Genomic Map