Assay Details
| Gene Symbol: Slc7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044740.2 | NP_001038205.1 | 7 - 8 | 1402 | 86 | |
| GenBank mRNA | AK154621.1 | - | 7 - 8 | 1376 | 86 | |
| AK155558.1 | - | 7 - 8 | 1378 | 86 | ||
| AK171369.1 | - | 7 - 8 | 1374 | 86 | ||
| CX212155.1 | - | 3 - 4 | 486 | 86 | ||
| M62838.1 | - | 7 - 8 | 1315 | 86 | ||
More Information
Set Membership: |
|

Slc7a2
Genomic Map