Assay Details
| Gene Symbol: Fam149a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153535.2 | NP_705763.1 | 1 - 2 | 1137 | 91 | |
| GenBank mRNA | AK140440.1 | - | 1 - 2 | 1137 | 91 | |
| AK141607.1 | - | 1 - 2 | 1080 | 91 | ||
| BC035537.1 | - | 1 - 2 | 1028 | 5369152 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam149a
Genomic Map