Assay Details
| Gene Symbol: Fam149a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153535.2 | NP_705763.1 | 5 - 6 | 1654 | 103 | |
| GenBank mRNA | AK034229.1 | - | 5 - 6 | 881 | 103 | |
| AK038552.1 | - | 5 - 6 | 869 | 103 | ||
| AK140440.1 | - | 5 - 6 | 1654 | 103 | ||
| AK141607.1 | - | 5 - 6 | 1597 | 103 | ||
| AK220518.1 | - | 5 - 6 | 890 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam149a
Genomic Map