Assay Details
| Gene Symbol: Fam149a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153535.2 | NP_705763.1 | 6 - 7 | 1826 | 93 | |
| GenBank mRNA | AK034229.1 | - | 6 - 7 | 1053 | 93 | |
| AK038552.1 | - | 6 - 7 | 1041 | 93 | ||
| AK140440.1 | - | 6 - 7 | 1826 | 93 | ||
| AK141607.1 | - | 6 - 7 | 1769 | 93 | ||
| AK220518.1 | - | 6 - 7 | 1062 | 93 | ||
| BC035537.1 | - | 6 - 7 | 1717 | 5369152 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam149a
Genomic Map