Assay Details
| Gene Symbol: Gnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010312.4 | NP_034442.1 | 5 - 6 | 454 | 68 | |
| GenBank mRNA | AK002604.1 | - | 5 - 6 | 449 | 68 | |
| AK146941.1 | - | 5 - 6 | 478 | 68 | ||
| AK151628.1 | - | 5 - 6 | 444 | 68 | ||
| BC029077.1 | - | 4 - 5 | 294 | 4019292 | 68 | |
| BC059942.1 | - | 5 - 6 | 480 | 6529685 | 68 | |
| BC062178.1 | - | 5 - 6 | 437 | 6330944 | 68 | |
| U34960.1 | - | 4 - 5 | 274 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnb2
Genomic Map