Assay Details
| Gene Symbol: Gnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010312.4 | NP_034442.1 | 9 - 10 | 1094 | 84 | |
| GenBank mRNA | AK002604.1 | - | 9 - 10 | 1090 | 84 | |
| AK146941.1 | - | 9 - 10 | 1118 | 84 | ||
| AK151628.1 | - | 9 - 10 | 1084 | 84 | ||
| BC029077.1 | - | 8 - 9 | 934 | 4019292 | 84 | |
| BC059942.1 | - | 9 - 10 | 1120 | 6529685 | 84 | |
| BC062178.1 | - | 9 - 10 | 1077 | 6330944 | 84 | |
| U34960.1 | - | 8 - 9 | 914 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126249 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126244 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126241 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126251 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126243 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126246 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126242 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126252 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126253 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126245 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126250 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987264 | BC059942.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987264 | BC062178.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987264 | AK002604.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987264 | AK146941.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987264 | BC029077.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 960562 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 523496 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136479 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987264 | BC059942.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987264 | BC062178.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987264 | AK002604.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987264 | AK146941.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987264 | BC029077.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P140311 | BC059942.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P140311 | BC062178.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P140311 | U38505.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P140311 | AK002604.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P140311 | AK146941.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126257 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136479 | - | - |
| Whole Mouse Genome G4122A | A_51_P140311 | BC059942.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P140311 | BC062178.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P140311 | U38505.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P140311 | AK002604.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P140311 | AK146941.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138340 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136479 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04165831 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04165823 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04165819 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04165796 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04565929 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126250 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126244 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126249 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126242 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126251 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126243 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126241 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126252 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126253 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126245 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126278 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04165795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04165837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04971136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126261 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126280 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126219 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04414446 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04165807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04912168 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04436941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04165794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04414445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04165816 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10534851 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534849 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534846 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534845 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534848 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534843 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534844 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534850 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534852 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534847 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534853 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450623_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10534842 | - | - |
| Mouse Genome 430 2.0 Array | 1450623_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1450623_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14693_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14693_PM_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa16420_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500031356.mm.1 | NM_010312.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031356.mm.1 | NM_010312.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031356.mm.1 | TC0500003737.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500031358.mm.1 | NM_010312.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031358.mm.1 | NM_010312.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031358.mm.1 | TC0500003737.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500031355.mm.1 | NM_010312.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031355.mm.1 | NM_010312.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031355.mm.1 | TC0500003737.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500031357.mm.1 | NM_010312.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031357.mm.1 | NM_010312.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031357.mm.1 | TC0500003737.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450623_PM_at | - | - |
| Mouse Expression Set 430 | 1450623_at | - | - |
| Murine Genome U74v2 Set | 96911_PM_at | - | - |
| Murine Genome U74v2 Set | 96911_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450623_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450623_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450623_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450623_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2350053 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002350053 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221732 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221732 | - | - |
| mouse-6 v1.1 (Illumina) | scl014693.1_109-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2350053 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl014693.1_109-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2350053 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1221732 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221732 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl014693.1_109-S | - | - |

Gnb2
Genomic Map