| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500024889.mm.1 |
NM_016690.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024889.mm.1 |
NM_016690.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024889.mm.1 |
TC0500002828.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500024888.mm.1 |
NM_016690.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024888.mm.1 |
NM_016690.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024888.mm.1 |
TC0500002828.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500024887.mm.1 |
NM_016690.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024887.mm.1 |
NM_016690.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024887.mm.1 |
TC0500002828.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500024885.mm.1 |
NM_016690.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024885.mm.1 |
NM_016690.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500024885.mm.1 |
TC0500002828.mm.1 |
MTA TC |