Assay Details
| Gene Symbol: Mb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013593.3 | NP_038621.2 | 1 - 2 | 144 | 73 | |
| GenBank mRNA | BC025172.1 | - | 1 - 2 | 156 | 3979003 | 73 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC025172.1, NM_013593.3 transcripts.
Set Membership: |
|

Mb
Genomic Map