| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500013812.mm.1 |
NM_001081394.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013812.mm.1 |
NM_001081394.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013812.mm.1 |
NM_027854.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013812.mm.1 |
NM_027854.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013812.mm.1 |
TC0500001528.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500013811.mm.1 |
NM_027854.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013811.mm.1 |
NM_027854.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013811.mm.1 |
TC0500001528.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500013810.mm.1 |
NM_027854.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013810.mm.1 |
NM_027854.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500013810.mm.1 |
TC0500001528.mm.1 |
MTA TC |