Assay Details
| Gene Symbol: Tmem248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081394.1 | NP_001074863.1 | 2 - 3 | 354 | 87 | |
| NM_027854.1 | NP_082130.1 | 2 - 3 | 416 | 87 | ||
| GenBank mRNA | AK002297.1 | - | 2 - 3 | 354 | 87 | |
| AK142176.1 | - | 2 - 3 | 404 | 87 | ||
| AK171152.1 | - | 2 - 3 | 382 | 87 | ||
| AK171607.1 | - | 2 - 3 | 352 | 87 | ||
| CN526906.1 | - | 2 - 3 | 441 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem248
Genomic Map