Assay Details
| Gene Symbol: Tmem248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081394.1 | NP_001074863.1 | 3 - 4 | 640 | 71 | |
| NM_027854.1 | NP_082130.1 | 3 - 4 | 702 | 71 | ||
| GenBank mRNA | AK002297.1 | - | 3 - 4 | 640 | 71 | |
| AK142176.1 | - | 3 - 4 | 690 | 71 | ||
| AK171152.1 | - | 3 - 4 | 668 | 71 | ||
| BC033455.1 | - | 3 - 4 | 612 | 4018584 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem248
Genomic Map