Assay Details
| Gene Symbol: Ccz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177682.3 | NP_808350.1 | 7 - 8 | 730 | 122 | |
| GenBank mRNA | AK090008.1 | - | 7 - 8 | 744 | 122 | |
| AK146704.1 | - | 7 - 8 | 763 | 122 | ||
| AK150793.1 | - | 7 - 8 | 767 | 122 | ||
| AK152328.1 | - | 7 - 8 | 766 | 122 | ||
| AK152576.1 | - | 7 - 8 | 766 | 122 | ||
| AK153160.1 | - | 7 - 8 | 767 | 122 | ||
| AK213761.1 | - | 4 - 5 | 301 | 122 | ||
| BC048077.1 | - | 7 - 8 | 730 | 6308253 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccz1
Genomic Map