Assay Details
| Gene Symbol: Chfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289577.1 | NP_001276506.1 | 4 - 5 | 463 | 148 | |
| NM_001289578.1 | NP_001276507.1 | 4 - 5 | 463 | 148 | ||
| NM_001289579.1 | NP_001276508.1 | 5 - 6 | 536 | 148 | ||
| NM_001289580.1 | NP_001276509.1 | 5 - 6 | 633 | 148 | ||
| NM_172717.4 | NP_766305.2 | 4 - 5 | 463 | 148 | ||
| GenBank mRNA | AK048930.1 | - | 5 - 6 | 613 | 148 | |
| AK052473.1 | - | 4 - 5 | 418 | 148 | ||
| AK077629.1 | - | 4 - 5 | 435 | 148 | ||
| AK133154.1 | - | 5 - 6 | 518 | 148 | ||
| AK147892.1 | - | 4 - 5 | 438 | 148 | ||
| AK148436.1 | - | 4 - 5 | 434 | 148 | ||
| AK154038.1 | - | 4 - 5 | 445 | 148 | ||
| AK155295.1 | - | 4 - 5 | 435 | 148 | ||
| AK155525.1 | - | 4 - 5 | 435 | 148 | ||
| AK169182.1 | - | 5 - 6 | 489 | 148 | ||
| AK169193.1 | - | 4 - 5 | 418 | 148 | ||
| BC049792.1 | - | 4 - 5 | 417 | 6510480 | 148 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK133154.1, AK169182.1, NM_001289579.1, NM_001289580.1 transcripts.
Set Membership: |
|

Chfr
Genomic Map