Assay Details
| Gene Symbol: Fam222a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004180.1 | NP_001004180.1 | 1 - 2 | 174 | 98 | |
| GenBank mRNA | BC027694.1 | - | 2 - 3 | 98 | ||
| BC057022.1 | - | 1 - 2 | 174 | 6813454 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC057022.1, NM_001004180.1 transcripts.
Set Membership: |
|

Fam222a
Genomic Map