| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700007058.mm.1 |
NM_001163684.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007058.mm.1 |
NM_001163684.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007058.mm.1 |
NM_025533.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007058.mm.1 |
NM_025533.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007058.mm.1 |
TC0700000827.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007053.mm.1 |
NM_001163684.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007053.mm.1 |
NM_001163684.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007053.mm.1 |
NM_025533.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007053.mm.1 |
NM_025533.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007053.mm.1 |
TC0700000827.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007059.mm.1 |
NM_001163684.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007059.mm.1 |
NM_001163684.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007059.mm.1 |
NM_025533.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007059.mm.1 |
NM_025533.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007059.mm.1 |
TC0700000827.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007052.mm.1 |
NM_001163684.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007052.mm.1 |
NM_001163684.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007052.mm.1 |
NM_025533.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007052.mm.1 |
NM_025533.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007052.mm.1 |
TC0700000827.mm.1 |
MTA TC |