Assay Details
| Gene Symbol: Nosip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163684.1 | NP_001157156.1 | 3 - 4 | 286 | 61 | |
| NM_025533.3 | NP_079809.1 | 3 - 4 | 286 | 61 | ||
| GenBank mRNA | AK007962.1 | - | 3 - 4 | 235 | 61 | |
| AK010006.1 | - | 3 - 4 | 222 | 61 | ||
| BC089029.1 | - | 3 - 4 | 212 | 30624825 | 61 | |
| DV648837.1 | - | 3 - 4 | 286 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nosip
Genomic Map