Assay Details
| Gene Symbol: Klk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027097.1 | NP_081373.1 | 3 - 4 | 560 | 122 | |
| GenBank mRNA | AK009217.1 | - | 2 - 3 | 423 | 122 | |
| BC147442.1 | - | 3 - 4 | 560 | 9056084 | 122 | |
| BC147463.1 | - | 3 - 4 | 560 | 9056105 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klk12
Genomic Map