Assay Details
| Gene Symbol: Rnf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016698.2 | NP_057907.2 | 8 - 9 | 1739 | 79 | |
| GenBank mRNA | AB026621.1 | - | 8 - 9 | 1738 | 79 | |
| AK034953.1 | - | 8 - 9 | 1738 | 79 | ||
| AK129106.1 | - | 8 - 9 | 1727 | 79 | ||
| AK146728.1 | - | 8 - 9 | 1739 | 79 | ||
| AK149972.1 | - | 8 - 9 | 1740 | 79 | ||
| AK165617.1 | - | 8 - 9 | 1739 | 79 | ||
| AK168523.1 | - | 8 - 9 | 1734 | 79 | ||
| AK172065.1 | - | 8 - 9 | 1737 | 79 | ||
| AK194340.1 | - | 1 - 2 | 94 | 79 | ||
| AK197020.1 | - | 2 - 3 | 162 | 79 | ||
| AK205789.1 | - | 1 - 2 | 94 | 79 | ||
| AK213334.1 | - | 1 - 2 | 82 | 79 | ||
| BC010342.1 | - | 8 - 9 | 1448 | 3602969 | 79 | |
More Information
Set Membership: |
|

Rnf10
Genomic Map