Assay Details
| Gene Symbol: Upk3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175309.4 | NP_780518.1 | 3 - 4 | 770 | 81 | |
| GenBank mRNA | AK042067.1 | - | 3 - 4 | 566 | 81 | |
| AK052525.1 | - | 3 - 4 | 772 | 81 | ||
| AK157647.1 | - | 3 - 4 | 577 | 81 | ||
| AY233463.1 | - | 3 - 4 | 564 | 81 | ||
| BC061224.1 | - | 3 - 4 | 567 | 81 | ||
| BC120594.1 | - | 3 - 4 | 520 | 40129517 | 81 | |
| BC137742.1 | - | 3 - 4 | 520 | 8860761 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Upk3b
Genomic Map