Assay Details
| Gene Symbol: Cyp26a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007811.2 | NP_031837.2 | 5 - 6 | 1095 | 71 | |
| GenBank mRNA | AF115769.1 | - | 5 - 6 | 1019 | 71 | |
| BC012673.1 | - | 5 - 6 | 1055 | 4210893 | 71 | |
| Y12657.1 | - | 5 - 6 | 1049 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp26a1
Genomic Map